Learn More
Macherey-Nagel™ Polypropylene Reservoir Column
Shop All Macherey-Nagel Products
Click to view available options
For Use With (Equipment):
1, 3, 6 mL Chromabond Polypropylene Columns
15, 45, 70 mL Chromabond Polypropylene Columns
Quantity:
1 Pack
10 Pack
Unit Size:
Each
Pack of 10
Specifications
Specifications
| Column Capacity (Metric) | 70 mL |
| Column Type | Reservoir |
| For Use With (Equipment) | 1, 3, 6 mL Chromabond Polypropylene Columns |
| Type | Reservoir Column |
| Includes | Adapter |
| Quantity | 1 Pack |