Learn More
Masterflex™ Antimicrobial Reducing Connector Barbed Fittings
These single barb fittings offer natural silver (Ag) protection for the fitting in applications where microorganism growth is likely. Using the AgION™ technology the fittings inhibit the growth of microorganisms on the surface of the fitting.
Brand:  Masterflex™ 06368-93
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11730339
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Prevents the growth of bacteria, fungi, mold and mildew
  • Easily connect fluid paths without the use of tools
  • Meets EPA standards
  • NSF 51, 61 and FDA compliant
  • Silver protection is not temperature sensitive and remains effective over time
  • Silver treatment will not contaminate your sample
  • Slip flexible tubing or hose onto the barb to create a seal that typically holds pressures up to 30 psi (2 bar)
  • No leaks, flow restrictions or costly downtime
  • Sterilization by ethylene oxide (ETO) or gamma
Specifications
Specifications
Barbed Fitting
1/4 x 3/8 in. tubing I.D.
Prevents growth of bacteria, fungi, mold and mildew
Ethylene Oxide, Gamma
Light Blue
Polypropylene
10 Pack
0.144 in. orifice
Reducing connector
30 psi (2 bar)