Learn More
Brady™ Polypropylene Prinzing Ball Valve Lockout
Description
- The two halves of the device encompass the ball valve lever to secure it from inadvertent activation of the valve.
- Locks out pipes from 12.70 to 31.75 mm in open and 12.70 to 63.50 mm closed position.
Specifications
Specifications
| Product Type | Valve Lockout |
| Alternate Part Number | BS08A-BL |
| Color | Blue |
| Material | Polypropylene |
| Certifications/Compliance | OSHA 1910.147 |
| Description | Prinzing Ball Valve Lockout |
| Dimensions (D x W x H) | 118.11 x 304.80 x 219.96 mm |
| Dimensions (L x W) | 305 x 220 mm |
| For Use With (Application) | Lockout Tagout |
| Height (English) | 8.66 in. |
| Show More |