Learn More
Brady™ Polypropylene Printer Labels
Product Code. 11777264
Change view
Click to view available options
:
Dimensions: 30W x 20mmH
Product Code.
11777264 Dimensions: 30W x 20mmH
1 options
Product Code. 11777264
Brand: Brady™ 114706
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815)
Description BMP71 Label Printer Labels
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Material Polypropylene
Print Technology Thermal Transfer
Product Type Printer Label
For Use With (Application) Voice and Data Communication
For Use With (Equipment) BMP™71
Labels per Row 1
Quantity 1 Ea.
Shape Rectangle
Size (Metric) 30W x 2 mmH
Temperature Range (Metric) -70°C to +80°C
Thickness (Metric) 0.028 mm
Vertical Repeat (Metric) 43.56 mm
Dimensions (LxW) 20 x 30 mm
Length (Metric) 15.24 m
Width (Metric) 30 mm
Height (Metric) 20.00 mm
Height (Metric) Printable Area 10.00 mm