Learn More
Cole-Parmer™ EPA Pre-Cleaned TOC Vial
Product Code. 12137770 Shop All Cole-Parmer Products
Change view
Click to view available options
Volume (Metric):
40 mL
Unit Size:
Pack of 80
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12137770 40 mL Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12137770 Supplier Cole-Parmer™ Supplier No. 0375620
  • ONLINE EXCLUSIVE PRICE
    5635.00 NOK
    5411.93 NOK / Pack of 80
    Save 223.07 NOK
    3% Off
Estimated Shipment: 27-11-2026
Use these vials for cleanroom applications. Cole-Parmer™ EPA Pre-Cleaned TOC Vials are manufactured from clear borosilicate class and include attached white polypropylene hole cap and PTFE/silicone septa.
  • Certified and traceable as associated with “Good Manufacturing Practices” (GMP)
  • Includes Certificate of Analysis
  • Packaged in an anti-static c-slotted plastic case
  • Cleaned and certified for Total Organic Carbon (TOC) to <10ppb.
Specifications
Material Borosilicate Glass
Volume (Metric) 40 mL
No. per Case 80 Pack