Learn More
BRAND™ Polypropylene Pouring Ring
Product Code. 15211259 Shop All Brand Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15211259 Polypropylene Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15211259 Supplier BRAND™ Supplier No. 122662
Estimated Shipment: 17-12-2026
Polypropylene Pouring Ring designed for use with laboratory bottles GL 32.
  • For use with laboratory bottles
  • Constructed of polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
For Use With (Equipment) GL 32 Laboratory Bottles
Product Type Pouring Ring