Learn More
Kartell™ Polypropylene Pipette Tips
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Pipette Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
Brand:  Kartell™ 0964800
 View more versions of this product
Product Code. 10469371
  • 1345.00 NOK / Pack of 200
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Description
Pluripet, Gilson, Socorex, polypropylene, light blue, 1000-5000 μL, rack
Specifications
Specifications
1000 to 5000 μL
Autoclavable
Rack
Polypropylene
Light Blue
CE / IVD
Kartell™, Gilson™, Socorex™, Nichiryo™