Learn More
BRAND™ Polypropylene Pipette Stand
Product Code. 10383431 Shop All Brand Products
Change view
Click to view available options
Positions:
94
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions Unit Size
10383431 94 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10383431 Supplier BRAND™ Supplier No. 30900
  • ONLINE EXCLUSIVE PRICE
    897.00 NOK
    700.20 NOK / Pack of 1
    Save 196.80 NOK
    21% Off
Estimated Shipment: 24-11-2026
Rotary model, with additional rotary tip reset to protect the pipette tips. Suitable for all graduated and one-mark pipettes. Slightly curved stand base rim helps retain dripping liquids.
  • Polypropylene construction
  • Fits 94 pipettes of different sizes
  • Rotary model
  • Additional rotary tip reset protect the pipette tips
  • Suitable for all graduated and one-mark pipettes
  • Slightly curved stand base rim helps retain dripping liquids
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Positions 94
Quantity 1 Each
Height (Metric) 450 mm
Width (Metric) 230 mm