Learn More
Brady™ Polypropylene Operating Tool
Description
- Operating tool which opens and locks the Pro-Lock II™ Cable Lockout Device.
Specifications
Specifications
| Product Type | Operating Tool |
| Alternate Part Number | PRO-LOCK SPANNER PP RED |
| Color | Red |
| Dimensions (L x W) | 26 x 152 mm |
| Quantity | 1 Ea. |