Learn More
Globe Scientific Polypropylene Multi-use Test Tubes
Description
- Multi-use tubes
- Can withstand 3000rpm
- Autoclavable and freezable
Specifications
Specifications
| Color | Clear |
| Capacity (Metric) | 5 mL |
| Length (Metric) | 75 mm |
| Material | Polypropylene |
| Packaging | 1000/Bag |
| Autoclavable | Autoclavable |
| Diameter (Metric) | 12 mm |
| Product Type | Test Tube |