Learn More
Ratiolab™ Polypropylene Multi Purpose Cup
Product Code. 15303492
Change view
Click to view available options
Capacity (Metric):
60 mL
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15303492 60 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15303492 Supplier Ratiolab™ Supplier No. 5921176
  • ONLINE EXCLUSIVE PRICE
    1150.00 NOK
    733.39 NOK / Pack of 500
    Save 416.61 NOK
    36% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Specifications
Sterility Sterile
Quantity 500 Pk.
Capacity (Metric) 60 mL
Product Type Multipurpose
Material Polypropylene
For Use With (Application) For sampling, transport and storage of solid, pasty and liquid samples
For Use With (Equipment) For sampling, transport and storage of solid, pasty and liquid samples