Description
- Polypropylene (PP) construction
- Includes tamper-evident screw cap and self-standing base
- Sterile
- Graduated
Specifications
Specifications
| Cap Style | Screw Cap |
| Certifications/Compliance | DIN EN 285 |
| Closure Material | Polypropylene |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 2 mL |
| Color | Transparent |
| Material | Polypropylene |
| For Use With (Application) | Storage of medical supplies such as serums and blood samples as well as samples heating , centrifugation etc. |
| For Use With (Equipment) | For Centrifuge |
| Height (Metric) | 44mm |
| Show More |