Learn More
Ratiolab™ Polypropylene Microcentrifuge Tubes
Product Code. 10008740
Change view
Click to view available options
Color:
Natural
Transparent
Capacity (Metric):
1.5 mL
2 mL
Unit Size:
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Capacity (Metric) Unit Size
10008740 Natural 1.5 mL Pack of 1000
10630983 Natural 2 mL Pack of 1000
10145884 Transparent 2 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10008740 Supplier Ratiolab™ Supplier No. 5615000
  • ONLINE EXCLUSIVE PRICE
    168.00 NOK
    106.52 NOK / Pack of 1000
    Save 61.48 NOK
    36% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Use these tubes for a variety of centrifugation applications. Ratiolab™ Polypropylene Microcentrifuge Tubes are available in a variety of colors for easy identification.
  • Autoclavable with opened cap at 121°C for 20 min.
  • Natural color or different colors for comfortable identification
  • Tight sealing cap
  • Safe and easy to be opened and closed
Specifications
Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Natural
Material Polypropylene
No. per Pack 1000/Pk.
Packaging 1000/Pk.