Learn More
Hirschmann™ Class B Graduated Cylinders, Blue Graduation, PP
Click to view available options
Capacity (Metric):
100 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 12
Pack of 6
Description
- Acc. to DIN 12681/ISO 6706
- PP
- Raised scale
- Main point ring graduation
- Hexagonal base
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Certifications/Compliance | DIN 12 681/ISO 6706 |
| Class | B |
| Includes | Elevated Scale |
| Type | Graduated Measuring Cylinder |