Learn More
Hirschmann™ Class B Graduated Cylinders, Blue Graduation, PP
Product Code. 10537631
Click to view available options
Capacity (Metric):
100 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 12
Pack of 6
Product Code. 10537631
Brand: Hirschmann™ 7520193
Only null left
Add to basket
Add to basket
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann™ Class B Measuring Cylinders, raised scale are manufactured from PP.
  • Acc. to DIN 12681/ISO 6706
  • PMP
  • Raised scale
  • transparent
  • Main point ring graduation
  • Hexagonal base
  • Conformity certified
Specifications
Capacity (Metric) 1000 mL
Material PMP
Tolerance ±10 mL
Certifications/Compliance DIN 12681, ISO 6706
Class A
Diameter (Metric) 69 mm
Includes Elevated Scale
Graduations 10 mL
Shape Round
For Use With (Application) Precise Liquid Measurement and Fluid Transfer in the Laboratory
Scale Color Blue
Base Type Hexagonal
Height (Metric) 440 mm
Type Graduated Measuring Cylinder