Learn More
Cole-Parmer™ Polypropylene Manifolds
Product Code. 11785238 Shop All Masterflex Products
Change view
Click to view available options
Length (Metric):
79.7 mm
106.1 mm
155.9 mm
186.4 mm
238.3 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11785238 106.1 mm Each
11775238 79.7 mm Each
11755258 186.4 mm Each
11725258 155.9 mm Each
11765258 238.3 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11785238 Supplier Cole-Parmer™ Supplier No. 3152209
  • ONLINE EXCLUSIVE PRICE
    702.00 NOK
    611.66 NOK / Pack of 1
    Save 90.34 NOK
    12% Off
Estimated Shipment: 28-10-2026
Specifications
Type Manifold
Inlet 3/8 in.
Material Polypropylene
No. of Ports 4
Port Size (Metric) 5.08 mm
Length (Metric) 106.1 mm
Width (Metric) 28.9 mm
Height (Metric) 28.95 mm