Learn More
Cole-Parmer™ Polypropylene Manifolds
Specifications
Specifications
| Type | Manifold |
| Inlet | 3/8 in. |
| Material | Polypropylene |
| No. of Ports | 10 |
| Port Size (Metric) | 5.08 mm |
| Length (Metric) | 238.3 mm |
| Width (Metric) | 28.9 mm |
| Height (Metric) | 28.95 mm |