Learn More
Cole-Parmer™ Polypropylene Manifold, 8 outlets
Product Code. 11745258
Change view
Click to view available options
Tubing Size:
1/2 in.
3/8in
Unit Size:
Each
Product Code. Tubing Size unitSize
11745258 1/2 in. Each
11735258 3/8in Each
2 options
Product Code. 11745258
Brand: Cole-Parmer™ 3152223
Only null left
Add to basket
Add to basket
Expand your flow and line capacity
  • Constructed of polypropylene.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Material Polypropylene
Overall Length (English) 8.75 in.
Overall Length (Metric) 22.23 cm
Min. Operating Temperature (English) 32°F
Min. Operating Temperature (Metric) 0°C
Tubing Size 1/2 in.
Inlet 1/2 in. NPT(F)
Outlet Port 3/8 in. (NPT(F)
Width (Metric) 3.81 cm
Width (English) 1.5 in.
Height (Metric) 3.81 cm
Height (English) 1.5 in.
Maximum Pressure 150 psi
Max. Operating Temperature (English) 210°F
Max. Operating Temperature (Metric) 99°C
Product Type PP Manifold, 8 Outlets