Learn More
Cole-Parmer™ Elbow Adapter, Compression x NPT(M), Polypropylene
Product Code. 11810110
Click to view available options
:
25 Pack, Fits 1/4 in. OD tubing, 1/8 in. NPT
Product Code. 11810110
Brand: Cole-Parmer™ 0644420
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cole-Parmer™ Elbow Adapter, Compression x NPT(M), Polypropylene features thread and a compression connections for a more permanent elbow adapter fitting.
  • Sturdy threaded fittings are designed for more permanent installation.
  • Use with our rigid tubing for high-pressure applications.
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With 1/4 in. OD tubing, 1/8 in. NPT connection
Packaging Quantity 25 Pack
Sterilization Method Autoclave Only
Width (Metric) 30 mm
Color Black
Product Type Male Pipe Elbow Adapter