Learn More
Brady™ Polypropylene: ISO Safety Sign - Safety shower
Product Code. 15634347
Click to view available options
Width (Metric):
148 mm
300 mm
400 mm
Height (Metric):
148 mm
300 mm
400 mm
Unit Size:
Each
Product Code. 15634347
Brand: Brady™ 815717
Only null left
Add to basket
Add to basket
Sign compliant to ISO 7010.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC E012-148x148-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 148 mm
Thickness 1.501 mm
Width (Metric) 148 mm
Temperature Range (Metric) -40°C to 120°C
Shape Square
Languages Global
Color White on Green