Learn More
Brady™ Polypropylene: ISO Safety Sign - Fire ladder
Product Code. 15634387
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Width (Metric):
100 mm
148 mm
250 mm
315 mm
400 mm
Height (Metric):
100 mm
148 mm
250 mm
315 mm
400 mm
missing translation for 'unitSize'
Each
Product Code. Width (Metric) Height (Metric) unitSize
15634387 250 mm 250 mm Each
15614387 100 mm 100 mm Each
15624387 148 mm 148 mm Each
15644387 315 mm 315 mm Each
15654387 400 mm 400 mm Each
5 missing translation for 'options'
Product Code. 15634387
missing translation for 'mfr': Brady™ 817207
Only null left
Add to basket
Add to basket
Sign compliant to ISO 7010.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC F003-250x250-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 250 mm
Thickness 1.501 mm
Width (Metric) 250 mm
Temperature Range (Metric) -40°C to 120°C
Shape Square
Languages Global
Color White on Red