Learn More
Brady™ Polypropylene: ISO Safety Sign - Fire ladder
Product Code. 15624387
Change view
Click to view available options
Width (Metric):
100 mm
148 mm
250 mm
315 mm
400 mm
Height (Metric):
100 mm
148 mm
250 mm
315 mm
400 mm
Unit Size:
Each
Product Code. Width (Metric) Height (Metric) unitSize
15624387 148 mm 148 mm Each
15614387 100 mm 100 mm Each
15634387 250 mm 250 mm Each
15644387 315 mm 315 mm Each
15654387 400 mm 400 mm Each
5 options
Product Code. 15624387
Brand: Brady™ 817205
Only null left
Add to basket
Add to basket
Sign compliant to ISO 7010.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC F003-148x148-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 148 mm
Thickness 1.501 mm
Width (Metric) 148 mm
Temperature Range (Metric) -40°C to 120°C
Shape Square
Languages Global
Color White on Red