Learn More
Brady™ Polypropylene: ISO Safety Sign - Collection of fire-fighting equipment
Product Code. 15684397
Change view
Click to view available options
Height (Metric):
100 mm
148 mm
200 mm
250 mm
315 mm
400 mm
Width (Metric):
100 mm
148 mm
200 mm
250 mm
315 mm
400 mm
Unit Size:
Each
Product Code. Height (Metric) Width (Metric) unitSize
15684397 315 mm 315 mm Each
15644397 100 mm 100 mm Each
15654397 148 mm 148 mm Each
15664397 200 mm 200 mm Each
15674397 250 mm 250 mm Each
15694397 400 mm 400 mm Each
6 options
Product Code. 15684397
Brand: Brady™ 817368
Only null left
Add to basket
Add to basket
Sign compliant to ISO 7010.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC F004-315x315-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 315 mm
Thickness 1.501 mm
Width (Metric) 315 mm
Temperature Range (Metric) -40°C to 120°C
Shape Square
Languages Global
Color White on Red