Learn More
Kartell™ Plastilab™ Straight-Bored Stopcocks
Product Code. 12388249
Change view
Click to view available options
Plug Bore:
4.9 mm
7 mm
10.9 mm
12.7 mm
Closure Size:
Nominal O.D.: 10 mm
Nominal O.D.: 13 mm
Nominal O.D.: 15 mm
Nominal O.D.: 8 mm
Unit Size:
Each
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Bore Closure Size Unit Size
12388249 4.9 mm Nominal O.D.: 8 mm Each
10564471 7 mm Nominal O.D.: 10 mm Pack of 10
10091051 12.7 mm Nominal O.D.: 15 mm Pack of 10
10603282 10.9 mm Nominal O.D.: 13 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12388249 Supplier Kartell™ Supplier No. 0037400
  • ONLINE EXCLUSIVE PRICE
    16.00 NOK
    15.23 NOK / Pack of 1
    Save 0.77 NOK
    4% Off
Estimated Shipment: 08-12-2026
Designed for liquids and gases under low pressure or low vacuum.
  • HDPE body interfaces with polypropylene tap
  • Operating pressure 0.5 bar
Specifications
Plug Bore 4.9 mm
Material Polypropylene, HDPE
Closure Size Nominal O.D.: 8 mm