Learn More
Karl Hecht™ Polypropylene (Griffin) Beaker
Product Code. p-7115724
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
400 mL
600 mL
1000 mL
2000 mL
5000 mL
Unit Size:
Each
Product Code. 10013621
Brand: Karl Hecht™ 41935012
Only null left
Add to basket
Add to basket
Use this low form, high transparency beaker for daily laboratory needs. Karl Hecht™ Polypropylene (Griffin) Beaker is antistatic, autoclavable, and includes a rim, spout, and printed blue volumetric scale.
  • Polypropylene construction.
  • Low form design.
  • High transparency, antistatic material.
  • Rim and spout.
  • Printed blue scale.
  • Use at temperatures up to 135°C.
  • Autoclavable (121°C).
Specifications
Diameter (Metric) Inner 31mm
Capacity (Metric) 25 mL
Material Polypropylene
Height (Metric) 47mm
Increments 1mL
Temperature (Metric) Max. Operating 135°C
Width (Metric) 31mm
Autoclavable Autoclavable
Graduated Yes
Certifications/Compliance ISO 7056
Style Griffin, Low Form