Learn More
Fisherbrand™ Polypropylene Graduated Microcentrifuge Tubes
Product Code. p-8001421
Click to view available options
Color:
Blue
Unit Size:
Pack of 500
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 11568242
Brand: Fisherbrand™ T33110B
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Use these tubes for a variety of applications. Fisherbrand™ Polypropylene Graduated Microcentrifuge Tubes are ideal for use with PCR systems and centrifugation.
  • Polypropylene graduated tubes
  • Autoclavable
  • Available in a variety of colors for easy identification
  • Available with pick-up tab
Specifications
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Blue
Material Polypropylene
For Use With (Application) For PCR, Centrifugation
No. per Pack 500/Pk.
Graduated Yes
Closure Type Snap Cap