Learn More
Sharpsguard™ dani 22 Multi-Purpose Waste Container
Click to view available options
Capacity (Metric):
21.01 L
Unit Size:
Each
Description
- Should be disposed of in landfill sites and therefore is not suitable for contaminated items
- Rigid polypropylene construction and wall thickness
- Offers outstanding protection
- All have clear British Standard markings, user instructions and a visible fill line
Specifications
Specifications
| Capacity (Metric) | 21.01 L |
| Color | Orange |