Learn More
Cole-Parmer™ Polypropylene Gel Loading Tip
Product Code. 15257546 Shop All Cole-Parmer Products
Click to view available options
Volume (Metric):
1 to 200 μL
Unit Size:
Each
Product Code. 15257546
Brand: Cole-Parmer™ 2571310
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Volume (Metric) 1 to 200 μL
Color Natural
Autoclavable Autoclavable
For Use With (Application) For DNA sequencing
Tip Style Gel Loading