Learn More
Ark Plas Products™ Polypropylene Female Tee Luer Adapter
Description
- Natural polypropylene luers
- Feature barbs designed with no parting line for reliable, leak-free use with a broad variety of medical and laboratory instruments, including syringetips and needles or stopcocks and needles
- Molded in virgin, lot-traceable material
- Ark-Plas Luer fittings include a large selection of both male and female luers available in locking or slip tip variances, as well as locking rings, bulkhead luers for panel mounting, threaded luers, adapters and plugs
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 4.8 mm |
| Min. Operating Temperature (Metric) | -17°C |
| Packaging Quantity | 25/Pk. |
| Shape | T-shape |
| Color | Natural |
| Max. Operating Temperature (Metric) | 135°C |