Learn More
Masterflex™ Polypropylene Female Luer Bulkhead Fitting
Specifications
Specifications
| Material | Polypropylene |
| For Use With | Fits 1/4-28 UNF thread to 1/8 in. hose barb |
| For Use With (Application) | Ideal for a variety of applications, including syringes or small tubing |
| Packaging Quantity | 25 Pack |
| Product Type | Female Luer Bulkhead Fitting |