Learn More
Multiroir™ Polypropylene Stackable Drawer
Store and organized items in protective polypropylene with the Multiroir™ Polypropylene Stackable Drawer. Available in white or dark gray, drawers are stackable and have a drawer-sliding capability for ease of use and spacesaving convenience.
Brand: Multiroir™ 18307
Additional Details : Weight : 1.22400kg
Description
- Stackable
- Raised edges for stacking and sliding ease
- Tight fitting lid
Specifications
25 L | |
Polypropylene | |
Drawer | |
Raised Edges | |
320 mm |
Gray | |
Rectangular | |
1 Each | |
410 mm | |
220 mm |