Learn More
Multiroir™ Polypropylene Stackable Drawer
Product Code. 11788371
Change view
Click to view available options
Capacity (Metric):
15 L
25 L
Color:
Dark Gray
Gray
White
Unit Size:
Each
Pack of 4
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color Unit Size
11788371 15 L White Each
30487000 15 L Dark Gray Pack of 6
30486998 25 L Gray Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11788371 Supplier Multiroir™ Supplier No. 18302
Estimated Shipment: 07-12-2026
Store and organized items in protective polypropylene with the Multiroir™ Polypropylene Stackable Drawer. Available in white or dark gray, drawers are stackable and have a drawer-sliding capability for ease of use and spacesaving convenience.
  • Stackable
  • Raised edges for stacking and sliding ease
  • Tight fitting lid
Specifications
Capacity (Metric) 15 L
Color White
Material Polypropylene
Shape Rectangular
Product Type Drawer
Quantity 1 Each
Style Raised Edges
Length (Metric) 370 mm
Width (Metric) 263 mm
Height (Metric) 174 mm