Learn More
Tenak™ Polypropylene Cryoboxes
Product Code. 15210564 Shop All TENAK Products
Change view
Click to view available options
Color:
Blue
Neutral
Red
Yellow
Holds:
1.2 to 2 mL Cryogenic Vials
Centrifuge Tubes
Unit Size:
Pack of 36
Product Code. Color Holds unitSize
15210564 Blue 1.2 to 2 mL Cryogenic Vials Pack of 36
15250564 Yellow 1.2 to 2 mL Cryogenic Vials Pack of 36
12459350 Blue Centrifuge Tubes Pack of 36
12479350 Neutral Centrifuge Tubes Pack of 36
12489350 Red Centrifuge Tubes Pack of 36
5 options
Product Code. 15210564
Brand: Tenak™ TE22300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Color Blue
Material Polypropylene
Shape Square
Type Cryo Box
Holds 1.2 to 2 mL Cryogenic Vials
Quantity 36 Pack
Length (Metric) 130 mm
Width (Metric) 130 mm
Height (Metric) 50 mm
Weight (Metric) 0.08 kg