Specifications
| Color | Red |
| Material | Polypropylene |
| Shape | Square |
| Type | Cryo Box |
| Holds | Centrifuge Tubes |
| Quantity | 36 Pack |
| Length (Metric) | 130 mm |
| Width (Metric) | 130 mm |
| Height (Metric) | 50 mm |
| Weight (Metric) | 0.08 kg |