Learn More
Cole-Parmer™ Polypropylene Copolymer Lab Containers
Product Code. 15237299 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (Metric):
2000 mL
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15237299 2000 mL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15237299 Supplier Cole-Parmer™ Supplier No. 0672735
  • ONLINE EXCLUSIVE PRICE
    2900.00 NOK
    2655.68 NOK / Pack of 50
    Save 244.32 NOK
    8% Off
Estimated Shipment: 14-01-2027
Specifications
Quantity 50 Pk.
Capacity (Metric) 2000 mL
Color Translucent
Product Type Lab Container
Material PPCO
Disposable Single-use
Height (Metric) 152.4 mm