Learn More
Buerkle™ Polypropylene Reducing Hose Connectors
Product Code. 10642482 Shop All Bürkle Products
Change view
Click to view available options
For Use With:
4 to 8/12 to 16 mm Hose
4 to 8/8 to 12 mm Hose
8 to 12/12 to 16 mm Hose
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
10642482 4 to 8/8 to 12 mm Hose Pack of 10
10630143 4 to 8/12 to 16 mm Hose Pack of 10
10739792 8 to 12/12 to 16 mm Hose Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10642482 Supplier Buerkle™ Supplier No. 87030408
  • ONLINE EXCLUSIVE PRICE
    208.00 NOK
    204.06 NOK / Pack of 10
    Save 3.94 NOK
    1% Off
Estimated Shipment: 04-12-2026
Use Buerkle™ Polypropylene Reducing Hose Connectors to make a connections between larger and smaller hoses.
  • Polypropylene
  • Sterilizable
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Foodstuffs, RoHS
Autoclavable Autoclavable
Material Polypropylene
For Use With 4 to 8/8 to 12 mm Hose
Packaging Quantity 10/Pk.; 10/Scale
Color White
Product Type Reducing Hose Connector