Learn More
Cole-Parmer™ Polypropylene Complete Coupling Fitting
Product Code. 11765489 Shop All Masterflex Products
Change view
Click to view available options
Quantity:
10 Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11765489 10 Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11765489 Supplier Cole-Parmer™ Supplier No. 3130500
  • ONLINE EXCLUSIVE PRICE
    1875.00 NOK
    1617.23 NOK / Pack of 10
    Save 257.77 NOK
    13% Off
Estimated Shipment: 11-11-2026
Specifications
For Use With 1/4 in. tubing I.D.
Min. Operating Temperature (Metric) -40°C
Maximum Pressure 125 psi
Max. Operating Temperature (Metric) 121°C
Quantity 10 Pk.