Learn More
Buerkle™ Polypropylene Compact Spigot
Product Code. 11911461 Shop All Bürkle Products
Change view
Click to view available options
Fits Tubing ID:
19.05 mm/GL14
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fits Tubing ID Unit Size
11911461 19.05 mm/GL14 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11911461 Supplier Buerkle™ Supplier No. 05301000
Estimated Shipment: 06-11-2026
Saves space with compact design. Buerkle™ Polypropylene Compact Spigot for canisters and barrels is ideal for use within refrigerators and on racks.
A compact spigot for canisters and barrels ideal for use within refrigerators and on racks, etc. The short discharge does not protrude over the lower edge of the container, making it the perfect choice especially when space is very limited.
  • Space-saving design
  • Optimised discharge for even flow
  • With connecting union nut for vertical position of the stop cock
  • Tight-closing for complete seal
TRUSTED_SUSTAINABILITY
Specifications
Type Compact Spigot
Material Polypropylene
Fits Tubing ID 19.05 mm/GL14
For Use With (Equipment) Canisters, Barrels, Ideal Addition to Flachmann Jerry Cans