Learn More
Gravis Polypropylene Screw Caps
Click to view available options
Thread Size (Metric):
20 mm
30 mm
33 mm
35 mm
38 mm
58 mm
110 mm
Color:
Black
White
Unit Size:
Pack of 100
Pack of 3
Description
- Available with or without a butyl-PTFE liner
- Available in either white or black color
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Pack | 100Pack |
| Quantity | 100Pack |
| Thread Size (Metric) | 30 mm |
| Liner Material | Unlined |
| Product Type | Cap |