Learn More
VITLAB™ Polypropylene Buchner Funnels
Click to view available options
Capacity (Metric):
6000 mL
Diameter (Metric)Top:
240mm
Unit Size:
Each
Description
- Upper and lower parts are detachable for easy cleaning
- Two different volumes and filter diameters are available
Specifications
Specifications
| Diameter (Metric)Top | 240mm |
| Capacity (Metric) | 6000 mL |
| Material | Polypropylene |
| Height (Metric) | 350 mm |
| Autoclavable | Autoclavable |
| Description | Polypropylene filter |
| Quantity | 1/Pk. |