Learn More
Sampling Systems™ Polypropylene Braided Cord
Product Code. 15859351 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
10 m
25 m
50 m
Unit Size:
10m
25m
50m
Product Code. Length (Metric) unitSize
15859351 50 m 50m
15839351 10 m 10m
15849351 25 m 25m
3 options
Product Code. 15859351
Brand: Sampling Systems™ 5070P50
Only null left
Add to basket
Add to basket
Sampling Systems™ Polypropylene Braided Cord is used to help lower the bucket of the Enviro Dipper.
  • For use with an Enviro Dipper
  • Polypropylene construction
Specifications
Product Type Braided Cord
Material Polypropylene
For Use With (Equipment) Enviro Dipper
Length (Metric) 50 m