Learn More
Sampling Systems™ Polypropylene Braided Cord
Product Code. 15839351 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
10 m
25 m
50 m
Unit Size:
10m
25m
50m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15839351 10 m 10m
15849351 25 m 25m
15859351 50 m 50m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15839351 Supplier Sampling Systems™ Supplier No. 5070P10
Only null left
Add to basket
Add to basket
Sampling Systems™ Polypropylene Braided Cord is used to help lower the bucket of the Enviro Dipper.
  • For use with an Enviro Dipper
  • Polypropylene construction
Specifications
Product Type Braided Cord
Material Polypropylene
For Use With (Equipment) Enviro Dipper
Length (Metric) 10 m