Learn More
Vitlab™ Polypropylene Bowl
Product Code. 10438502
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
4 L
7 L
Unit Size:
Each
Pack of 3
Pack of 5
Product Code. Capacity (Metric) unitSize
10438502 1 L Each
10528341 2 L Each
10321742 2 L Pack of 5
10232222 1 L Pack of 5
10351022 7 L Pack of 3
10098680 4 L Each
10496871 4 L Pack of 5
7 options
Product Code. 10438502
Brand: Vitlab™ 42594
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these bowls to help prevent splashing on the work surface while working with strong reactions in uncovered vessels. VITLAB™ Polypropylene Bowls feature a broad, stable edge and circumferential standing ring on the bottom.
  • White round bowl
  • Polypropylene
Specifications
Capacity (Metric) 1 L
Material Polypropylene
Height (Metric) 70 mm
Diameter (Metric) 160 mm