Learn More
Hettich™ Polypropylene Bottle with Screw Cap
Product Code. 10019332
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10019332 1000 mL Each
1 options
Product Code. 10019332
Brand: Hettich™ 4239
Only null left
Add to basket
Add to basket
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Polypropylene Bottle with Screw Cap
  • Compatible with a variety of Hettich Rotors, Carriers and Adapters
Specifications
Capacity (Metric) 1000 mL
Material Polypropylene
Length (Metric) 176mm
Product Type Centrifuge Bottle
Diameter (Metric) 96 mm
Closure Type Screw Cap