Learn More
Buerkle™ Bottle Holder for Buerkle™ Telescopic Rods
Product Code. 10333921 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
750 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10333921 750 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10333921 Supplier Buerkle™ Supplier No. 53544100
Estimated Shipment: 17-11-2026
Holds all bottles up to 88 mm (dia.). Plastic, glass or metal bottles can be securely filled in this way. Angle steplessly adjustable up to 90°.
  • With universal quick tightening strap
  • Polypropylene/PA
  • For industry and water treatment
TRUSTED_SUSTAINABILITY
Specifications
Product Type Water Sampler
Material Polypropylene, Polyamide
Includes Universal quick tightening strap
Capacity (Metric) 750 mL