Learn More
Brady™ Polypropylene Blank Heavy Duty Floor Stand
Product Code. 10011304 Shop All Brady Products
Change view
Click to view available options
Color:
Yellow
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10011304 Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10011304 Supplier Brady™ Supplier No. 092110
  • ONLINE EXCLUSIVE PRICE
    752.00 NOK
    738.60 NOK / Pack of 1
    Save 13.40 NOK
    1% Off
View all promotions
In Stock
Easily transportable, the Polypropylene Blank Heavy Duty Floor Stand is very useful to carry with and clearly indicate substantial dangers such as slippery zones. It is ideal for hospitals or public halls for example.
  • Made of polypropylene, our maintenance-free stands will not dent.
  • Printable area is 160 mm x 200 mm.
Specifications
Alternate Part Number HEAVY DUTY FLOORSTAND -YELLOW-BLANK-AUST
Displayed Text Blank
Height (English) 25.98 in.
Height (Metric) 660 mm
Packaging Quantity 1 Ea.
Shape Rectangle
Color Yellow