Learn More
Lenz Laborglasinstrumente™ Polypropylene Bent Hose Connectors
Formulated from polypropylene. Lenz Laborglasinstrumente™ Polypropylene Bent Hose Connectors are for use with screw caps with bore.
161.12 NOK - 778.04 NOK
Specifications
Product Type | Bent Hose Connection |
---|---|
Autoclavable | Autoclavable |
Material | Polypropylene |
For Use With | For screw caps with bore |
Packaging Quantity | 10 Pack |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
12323379
|
Lenz Laborglasinstrumente™
01360001 |
GL18 |
778.04 NOK
Pack of 10 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
12313379
|
Lenz Laborglasinstrumente™
01360000 |
GL14 |
161.12 NOK
Pack of 10 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
- For screw caps with bore
- Polypropylene construction
Specifications
Bent Hose Connection | |
Polypropylene | |
10 Pack |
Autoclavable | |
For screw caps with bore | |
Bent |