Learn More
Semadeni™ Color-Coded Plastic Beakers
Description
- Translucent colors
- 30mL capacity
- Graduated
- Optional LDPE lids available for purchase
Specifications
Specifications
| Color | Natural |
| Capacity (Metric) | 30 mL |
| Material | Polypropylene |
| For Use With (Application) | Suitable as Dosing or Laboratory Beakers |
| Quantity | 100 Pk. |
| Type | Beaker |
| Disposable | Single-use |
| Graduated | Yes |