Learn More
Semadeni™ Color-Coded Plastic Beakers
Product Code. 11767192
Click to view available options
Color:
Blue
Natural
Yellow
Unit Size:
Pack of 100
Product Code. 11767192
Brand: Semadeni™ 8612
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Excellent for the storage, mixing or pouring of liquids in the laboratory. Semadeni™ Color-Coded Plastic Beakers are manufactured from durable polypropylene.
  • Translucent colors
  • 30mL capacity
  • Graduated
  • Optional LDPE lids available for purchase
Specifications
Color Blue
Capacity (Metric) 30 mL
Material Polypropylene
For Use With (Application) Suitable as Dosing or Laboratory Beakers
Quantity 100 Pk.
Type Beaker
Disposable Single-use
Graduated Yes