Description
- Barrel screw joint made of PP for secure connection in the barrel in three different thread models
- For drawing off liquids that are not flammable
- The container connectors make decanting even easier and safer with their more secure connection to barrels
- The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange
Specifications
Specifications
| For Use With | PTFE Barrel Pump |
| Material | Polypropylene |
| Size | Tri-Sure, Coarse Outer Thread |
| Type | Barrel Screw Joint |